Sample Bags without Closure
800×600 mm, 0,05 mm
Catalog # SCAC2046
Supplier: SCANCELL FOLIEN VETRIEB
Jump to:
Product Details & Documents
Bags made from PE transparent, for the storage or transport of samples.
Available in several sizes.
Caution: Supplied without closure ties.
Documents
SCAC2046_TDS_EN SCAC20771_TDS_EN
Datasheet
Specifications
- L×B:800×600 mm
- Seq:15
- Tykkelse:0,05 mm