Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect column to cooler (50-150 mm column)
Catalog # HEWLG1316-87321
Capillaries for LC systems
Catalog # HEWLG1316-87321
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect column to cooler (50-150 mm column)
Price
717,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Beskrivelse:
    Capillaries, non-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect column to cooler (50-150 mm column)
  • Seq:
    39