Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, PP, Metal foil
Katalog # 548-2047
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
- Clarity of glass
- Can be safely incinerated
- Lightweight and virtually unbreakable
* With cap on
Specifikationer
- Kasket inkluderet:Yes
- Hættemateriale:PP
- Diameter×H:27×61* mm
- Septum:Metal foil
- Kapacitet:20,00 ml