Kolonitæller, eCount™
Electronic colony counter
Katalog # 710-0596
Leverandør: HEATHROW
Gå til:
Product Details & Documents
The all-in-one eCount™ combines a multifunctional electronic counter and a Sharpie® pen for marking Petri dishes to prevent missing or double-counting colonies.
- Five-digit display
- Touch controls for multiple counts: Up to 32 different zones
- Counter automatically beeps with each count, or select the LED flash mode for a visual alert of each count
- Working temperature from 0 to 50 °C
- Light ergonomic body balanced for easy manipulation
Designed for rapid, accurate counts. Press the lock key to write with the Sharpie® pen without counting. The Sharpie® pen provides highly visible marks on both plastic and glass dishes. Counter accepts both fine and extra fine point Sharpie® pens. Easily change colours for separate counts.
Delivery information: Includes a three-volt CR1225 lithium battery, one fine-point Sharpie® pen and weighted stand. Five years warranty, battery not included in warranty.
Specifikationer
- Beskrivelse:Electronic colony counter