Orders LinkContactUsLinkComponent
Accessories for temperature control systems, Integral XT
Accessories for temperature control systems, Integral XT
Double connector, M30×1,5 outer thread to NPT 3/4" outer thread
Katalog # LAUDEOV207
Leverandør:   LAUDA
Accessories for temperature control systems, Integral XT
Katalog # LAUDEOV207
Leverandør:  LAUDA
Accessories for temperature control systems, Integral XT
Accessories for temperature control systems, Integral XT
Double connector, M30×1,5 outer thread to NPT 3/4" outer thread
Price
774,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Double connector, M30×1,5 outer thread to NPT 3/4" outer thread
  • Seq:
    09