Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,5 ± ml
Katalog # 612-3212
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
- Marks and inscriptions in high contrast blue enamel
- Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
- NS:29/32
- Kapacitet:2000 ml
- Seq:12
- Tolerance:0,5 ± ml