Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,5 ± ml
Katalog # 612-3212
Leverandør:   SciLabware
Volumetric flasks, class A, blue markings
Katalog # 612-3212
Leverandør:  SciLabware
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,5 ± ml
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
  • NS:
    29/32
  • Kapacitet:
    2000 ml
  • Seq:
    12
  • Tolerance:
    0,5 ± ml