Volumetric flasks, class A, white markings
2000 ml, 29/32, 0,50 ± ml
Katalog # 612-3245
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- White enamel inscriptions, ideal for use with light sensitive solutions
: BS EN ISO 1042, DIN 12664
Specifikationer
- NS:29/32
- Kapacitet:2000 ml
- Seq:11
- Tolerance:0,50 ± ml