Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
500 ml, 19/26, 0,20 ± ml
  612-3242
 :   SciLabware
Volumetric flasks, class A, white markings
  612-3242
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
500 ml, 19/26, 0,20 ± ml
828,00 kr
1 * 2 Items
PYREX®, borosilicate glass, amber, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • White enamel inscriptions, ideal for use with light sensitive solutions
 : BS EN ISO 1042, DIN 12664
  • NS:
    19/26
  • Kapacitet:
    500 ml
  • Seq:
    09
  • Tolerance:
    0,20 ± ml