Volumetric flasks, class A, blue markings
250 ml, 14/23, 0,12 ± ml
612-3209
PYREX®, borosilicate glass, amber, with polyethylene stopper.
- Marks and inscriptions in high contrast blue enamel
- Batch coded for traceability and supplied with a batch certificate
: ISO 1042, DIN 12 664
- NS:14/23
- Kapacitet:250 ml
- Seq:09
- Tolerance:0,12 ± ml