Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
250 ml, 14/23, 0,12 ± ml
  612-3209
 :   SciLabware
Volumetric flasks, class A, blue markings
  612-3209
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
250 ml, 14/23, 0,12 ± ml
1.240,00 kr
1 * 2 Items
PYREX®, borosilicate glass, amber, with polyethylene stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Batch coded for traceability and supplied with a batch certificate
 : ISO 1042, DIN 12 664
  • NS:
    14/23
  • Kapacitet:
    250 ml
  • Seq:
    09
  • Tolerance:
    0,12 ± ml