Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,1 ± ml
Katalog # 612-3208
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
- Marks and inscriptions in high contrast blue enamel
- Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
- NS:14/23
- Kapacitet:200 ml
- Seq:08
- Tolerance:0,1 ± ml