Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 14/23, 0,08 ± ml
Katalog # 612-3207
Leverandør:   SciLabware
Volumetric flasks, class A, blue markings
Katalog # 612-3207
Leverandør:  SciLabware
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 14/23, 0,08 ± ml
Price
2.010,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
  • NS:
    14/23
  • Kapacitet:
    100 ml
  • Seq:
    07
  • Tolerance:
    0,08 ± ml