Volumetric flasks, class A, blue markings
100 ml, 14/23, 0,08 ± ml
Katalog # 612-3207
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
- Marks and inscriptions in high contrast blue enamel
- Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
- NS:14/23
- Kapacitet:100 ml
- Seq:07
- Tolerance:0,08 ± ml