Volumetric flasks, class A, blue markings
50 ml, 12/21, 0,05 ± ml
Katalog # 612-3206
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with polyethylene stopper.
- Marks and inscriptions in high contrast blue enamel
- Batch coded for traceability and supplied with a batch certificate
Certifications: ISO 1042, DIN 12 664
Specifikationer
- NS:12/21
- Kapacitet:50 ml
- Seq:05
- Tolerance:0,05 ± ml