Orders LinkContactUsLinkComponent
Nessler cylinders
Nessler cylinders
Nessler cylinder with marking at 50 ml and 100 ml, 180 mm height
Katalog # BIBBCRC322
Leverandør:   SciLabware
Nessler cylinders
Katalog # BIBBCRC322
Leverandør:  SciLabware
Nessler cylinders
Nessler cylinders
Nessler cylinder with marking at 50 ml and 100 ml, 180 mm height
Price
392,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc
  • Made of specially selected borosilicate glass tubing
  • Supplied in matched pairs for uniformity
Specifikationer
  • Beskrivelse:
    Nessler cylinder with marking at 50 ml and 100 ml, 180 mm height
  • Seq:
    02