Volumetric flasks, class A, white markings
1000 ml, 24/29, 0,30 ± ml
Katalog # 612-3247
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, amber, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- White enamel inscriptions, ideal for use with light sensitive solutions
Certifications: BS EN ISO 1042, DIN 12664
Specifikationer
- NS:24/29
- Kapacitet:1000 ml
- Seq:10
- Tolerance:0,30 ± ml