Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
50 ml, 12/21, 0,05 ± ml
Catalog # 612-3251
Supplier:   SciLabware
Volumetric flasks, class A, white markings
Catalog # 612-3251
Supplier:  SciLabware
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
50 ml, 12/21, 0,05 ± ml
Price
1.090,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass, amber, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • White enamel inscriptions, ideal for use with light sensitive solutions
Certifications: BS EN ISO 1042, DIN 12664
Specifications
  • NS:
    12/21
  • Kapacitet:
    50 ml
  • Seq:
    04
  • Tolerance:
    0,05 ± ml