Volumetric flasks, class A, white markings
50 ml, 12/21, 0,05 ± ml
Catalog # 612-3251
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, amber, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- White enamel inscriptions, ideal for use with light sensitive solutions
Certifications: BS EN ISO 1042, DIN 12664
Specifications
- NS:12/21
- Kapacitet:50 ml
- Seq:04
- Tolerance:0,05 ± ml