Volumetric flasks, class A, white markings
500 ml, 19/26, 0,20 ± ml
612-3198
PYREX®, borosilicate glass, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- Calibrated to contain (TC, In)
- White enamel inscriptions
Certifications: BS EN ISO 1042, DIN 12664
- NS:19/26
- Kapacitet:500 ml
- Tolerance:0,20 ± ml