Volumetric flasks, class A, white markings
20 ml, 10/19, 0,03 ± ml
Katalog # 612-2902
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- Calibrated to contain (TC, In)
- White enamel inscriptions
Certifications: BS EN ISO 1042, DIN 12664
Documents
Specifikationer
- NS:10/19
- Kapacitet:20 ml
- Tolerance:0,03 ± ml