Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
5 ml, 10/19, 0,02 ± ml
Katalog # 612-3233
Leverandør:   SciLabware
Volumetric flasks, class A, white markings
Katalog # 612-3233
Leverandør:  SciLabware
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
5 ml, 10/19, 0,02 ± ml
Price
555,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
  • White enamel inscriptions
Certifications: BS EN ISO 1042, DIN 12664
Specifikationer
  • NS:
    10/19
  • Kapacitet:
    5 ml
  • Tolerance:
    0,02 ± ml