Centrifugerør, Axygen®
15 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack
Katalog # 525-1746
Leverandør: Corning
Gå til:
Product Details & Documents
PP, conical, printed graduation, with HDPE flat screw caps.
- Max. RCF: 12 000×g
- Easy one hand operation
- Large writing area for sample identification
Specifikationer
- Lukkestørrelse:Skruelåg
- Pakket:25 tubes and assembled caps per rack
- Version:Sterile
- Kapacitet:15 ml
- Seq:03