Orders LinkContactUsLinkComponent
Centrifugerør, Axygen®
15 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack
Katalog # 525-1746
Leverandør:   Corning
Centrifugerør, Axygen®
Katalog # 525-1746
Leverandør:  Corning
Centrifugerør, Axygen®
Centrifugerør, Axygen®
15 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack
Price
1.730,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
PP, conical, printed graduation, with HDPE flat screw caps.
  • Max. RCF: 12 000×g
  • Easy one hand operation
  • Large writing area for sample identification
Specifikationer
  • Lukkestørrelse:
    Skruelåg
  • Pakket:
    25 tubes and assembled caps per rack
  • Version:
    Sterile
  • Kapacitet:
    15 ml
  • Seq:
    03