Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Magnetic stirring bars, PTFE, triangular, BOLA

40 mm, 14 mm, store beholdere
Katalog # BOHLC357-18
Leverandør:   Bohlender
Magnetic stirring bars, PTFE, triangular, BOLA
Katalog # BOHLC357-18
Leverandør:  Bohlender
Magnetic stirring bars, PTFE, triangular, BOLA
Magnetic stirring bars, PTFE, triangular, BOLA

40 mm, 14 mm, store beholdere

Price
88,20 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Triangular shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE.

  • Chemical resistant

Temperature resistance: −200 to +250 °C.

Most commonly used for big vessels in which strong turbulence can be acheived with low speeds. These can also been useful for mixing of reagents which resist dissolving and for avoiding any residues at the bottom of the vessels.

Specifikationer
  • Ø:
    14 mm
  • Til:
    store beholdere
  • Længde:
    40 mm
  • Seq:
    06