Magnetic stirring bars, PTFE, triangular, BOLA
20 mm, 8 mm, store beholdere
Katalog # BOHLC357-06
Leverandør: Bohlender
Gå til:
Product Details & Documents
Triangular shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE.
- Chemical resistant
Temperature resistance: −200 to +250 °C.
Most commonly used for big vessels in which strong turbulence can be acheived with low speeds. These can also been useful for mixing of reagents which resist dissolving and for avoiding any residues at the bottom of the vessels.
Specifikationer
- Ø:8 mm
- Til:store beholdere
- Længde:20 mm
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



