Orders LinkContactUsLinkComponent
Test kits, DEHA, CHECKIT®
DEHA test kit, 0 - 0,5 mg/L DEHA, 1 KIT
Katalog # LOVI147370
Leverandør:   Lovibond
Test kits, DEHA, CHECKIT®
Katalog # LOVI147370
Leverandør:  Lovibond
CAS nummer:  
Test kits, DEHA, CHECKIT®
DEHA test kit, 0 - 0,5 mg/L DEHA, 1 KIT
CAS nummer:  
Price
910,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
 : Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    DEHA test kit
  • Område:
    0 - 0,5 mg/L DEHA
Specifikationer
Accessories
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • CHECKIT® Comparator, path length 13,5 mm
    CHECKIT® Comparator, path length 13,5 mm
    LOVI145000
    1
    Log ind for tilgængelighed
    1 * 1 Items - 218,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    LOVI145500
    10
    Log ind for tilgængelighed
    1 * 10 Items - 493,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    LOVI145505
    5
    Log ind for tilgængelighed
    1 * 5 Items - 292,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    LOVI145510
    100
    Log ind for tilgængelighed
    1 * 100 Items - 4.080,00 kr