Orders LinkContactUsLinkComponent
Cover spectacles, Squale
Cover spectacles, Squale
UV-filter, dugfri, ridsefast, transparent/sort, Clear PC
Katalog # 111-0387
Leverandør:   Bollé Safety
Cover spectacles, Squale
Katalog # 111-0387
Leverandør:  Bollé Safety
Cover spectacles, Squale
Cover spectacles, Squale
UV-filter, dugfri, ridsefast, transparent/sort, Clear PC
Price
77,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cover glasses (OTG) give excellent protection for wearers of prescription eyewear. Supplied with adjustable cord.
  • Længdejusterbare nylonstænger
  • Formstøbt linse, pandebeskyttelse og sideskjolda
  • Indirekte ventilation
Vægt: 43 g
 : EN 166, EN 170
Specifikationer
  • Model:
    UV-filter, dugfri, ridsefast
  • Farve:
    transparent/sort
  • Linser:
    Clear PC