Orders LinkContactUsLinkComponent
Accessories for RapidVap®
Accessories for RapidVap®
Radioisotope trap insert with molecular sieves for protecting the rotary vane vacuum pump (requires a clear PP canister with mounting bracket or stand for proper use)
Katalog # 531-0216
Leverandør:   LABCONCO
Accessories for RapidVap®
Katalog # 531-0216
Leverandør:  LABCONCO
Accessories for RapidVap®
Accessories for RapidVap®
Radioisotope trap insert with molecular sieves for protecting the rotary vane vacuum pump (requires a clear PP canister with mounting bracket or stand for proper use)
Price
1.470,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Radioisotope trap insert with molecular sieves for protecting the rotary vane vacuum pump (requires a clear PP canister with mounting bracket or stand for proper use)
  • Seq:
    17