Cellulose, microcrystalline
Glass bottle for solids, 500 g
CAS No.: 9004-34-6 Physical Form: Powder Appearance: White Merck Reference: 14,1965 MDL No.: MFCD00081512
: Microcrystallinecellulose
- :9004-34-6
- :(C₁₂H₂₂O₁₁)n
- :MFCD00081512
- :342.3 g/mol
- :14,1965
- :260…270 °C
- :1,27…1.6 g/cm³ (20 °C)
- :Stue temperatur
- :232-674-9
- Pakning:Glass bottle for solids
- Pakn:500 g



