Orders LinkContactUsLinkComponent
Mortars with pestles
Mortars with pestles
1000 ml, 105 mm, 92 mm, 180 mm, 190 mm
Katalog # 410-0106
Leverandør:   USBECK Laborgeräte
Mortars with pestles
Katalog # 410-0106
Leverandør:  USBECK Laborgeräte
Mortars with pestles
Mortars with pestles
1000 ml, 105 mm, 92 mm, 180 mm, 190 mm
Price
1.910,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Mortar and pestle made of stainless steel.
  • Slip-resistant feet
Hand wash only.
Specifikationer
  • Dybde:
    92 mm
  • Ø:
    190 mm
  • Ø indv.:
    180 mm
  • Kapacitet:
    1000 ml
  • Højde:
    105 mm