Containers, rectangular
12,5 l, 395×300×165 mm, hvid, HDPE
Katalog # SEMA1296
Leverandør: SEMADENI
Gå til:
Product Details & Documents
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.
Specifikationer
- Kapacitet:12,5 l
- Farve:hvid
- Materiale:HDPE
- Seq:03
- B×D×H:395×300×165 mm