Vacuum Controller, V-850/V-855
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Leveringsinformation: Delivered with an air vent valve and pressure sensors.
Specifikationer
- Beskrivelse:Vakuumstyreenhed V-855 til R-200/R-205
- Seq:03
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Kommunikationskabel til R-210/R-215
Produktinformation
- Katalog #TilgængelighedPrice
- Woulff-flaske til V-850/V-855
Produktinformation
- Katalog #TilgængelighedPrice
- Automatisk destillationsprobe til V-855Automatisk destillationsprobe til V-855531-0181Log ind for tilgængelighed1 * 1 Items - 2.090,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Monteringssæt til R-210/R-215
Produktinformation
- Katalog #TilgængelighedPrice
- Fjernstyring til V-850/V-855