Orders LinkContactUsLinkComponent
Instrument dishes
Låg til instrumentbakker 1000/1800 ml, PS krystalklar, 300×170×20 mm
  BURK4230-2101
 :   BURKLE
Instrument dishes
  BURK4230-2101
 :  BURKLE
Instrument dishes
Instrument dishes
Låg til instrumentbakker 1000/1800 ml, PS krystalklar, 300×170×20 mm
159,00 kr
1 * 1 Items
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
  • Melamine, white
  • Tray without lid
Lid available as accessory.
  • Beskrivelse:
    Låg til instrumentbakker 1000/1800 ml, PS krystalklar
  • B×D×H:
    300×170×20 mm