Milk butyrometer
Butyrometer for milk, 0 - 6% : 0,1
Catalog # 709-0142
Supplier: GERBER FUNKE
Jump to:
Product Details & Documents
Borosilicate glass.
- With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specifications
- Beskrivelse:Butyrometer for milk, 0 - 6% : 0,1
- Seq:01
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 614-2028Stopper, rubber, conical, for all measuring method butyrometersLog in for availability1 * 100 Items - 360,00 kr