Electrode Cables
Electrode cable, with BNC plug, for connecting electrodes with Metrohm plug-in head G with a measuring instrument, 1 m
Katalog # METR6.2104.090
Leverandør: METROHM
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Electrode cable, with BNC plug, for connecting electrodes with Metrohm plug-in head G with a measuring instrument
- Længde:1 m
- Seq:11