Orders LinkContactUsLinkComponent
PT1000 precision probes
PT1000 precision probes
Pointed PT1000 probe with 600 mm red silicone cable, TPX 400, 3×120 mm
Katalog # 620-1399
Leverandør:   EBRO
PT1000 precision probes
Katalog # 620-1399
Leverandør:  EBRO
PT1000 precision probes
PT1000 precision probes
Pointed PT1000 probe with 600 mm red silicone cable, TPX 400, 3×120 mm
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Pointed PT1000 probe with 600 mm red silicone cable, TPX 400
  • Seq:
    08
  • Ø×L:
    3×120 mm