Orders LinkContactUsLinkComponent
Accessories for pH electrodes
Accessories for pH electrodes
Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
Katalog # WTWA109735
Leverandør:   WTW
Accessories for pH electrodes
Katalog # WTWA109735
Leverandør:  WTW
Accessories for pH electrodes
Accessories for pH electrodes
Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
Price
538,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
  • Seq:
    06