Orders LinkContactUsLinkComponent
Accessories for RapidVap®
Accessories for RapidVap®
Vacuum tubing kit, two each 12,7 mm ID vacuum tubing (122 cm length) and 4 tubing clamps
Katalog # LBCP7814500
Leverandør:   LABCONCO
Accessories for RapidVap®
Katalog # LBCP7814500
Leverandør:  LABCONCO
Accessories for RapidVap®
Accessories for RapidVap®
Vacuum tubing kit, two each 12,7 mm ID vacuum tubing (122 cm length) and 4 tubing clamps
Price
1.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Vacuum tubing kit, two each 12,7 mm ID vacuum tubing (122 cm length) and 4 tubing clamps
  • Seq:
    18