Bottle top tilt measures
2 ml, klar
613-6253
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32
- Kapacitet:2 ml
- Farve:klar