Orders LinkContactUsLinkComponent
Bottle top tilt measures
2 ml, klar
  613-6253
Bottle top tilt measures
  613-6253
Bottle top tilt measures
Bottle top tilt measures
2 ml, klar
485,00 kr
1 * 1 Items
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32
  • Kapacitet:
    2 ml
  • Farve:
    klar