Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
Katalog # REST24324
Leverandør:   Restek
Glass wool
Katalog # REST24324
Leverandør:  Restek
Glass wool
Glass wool
Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
Price
448,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
  • Seq:
    01