Orders LinkContactUsLinkComponent
Safety funnels
Safety funnels
180 mm påfyldningsåbning, S 56×4
Katalog # DUEH5.10.007
Leverandør:   DUPERTHAL
Safety funnels
Katalog # DUEH5.10.007
Leverandør:  DUPERTHAL
Safety funnels
Safety funnels
180 mm påfyldningsåbning, S 56×4
Price
2.590,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel safety funnels with hinged lid and butress thread S 56×4.
  • Overfilling protection
  • Lid reduces the escaping vapours
  • Flame arrester made from stainless steel (simple to interchange)
Specifikationer
  • Gevind:
    S 56×4
  • Beskrivelse:
    180 mm påfyldningsåbning