Orders LinkContactUsLinkComponent
Safety funnels
120 mm påfyldningsåbning, S 56×4
  DUEH5.10.001
 :   DUPERTHAL
Safety funnels
  DUEH5.10.001
 :  DUPERTHAL
Safety funnels
Safety funnels
120 mm påfyldningsåbning, S 56×4
1.580,00 kr
1 * 1 Items
Stainless steel safety funnels with hinged lid and butress thread S 56×4.
  • Overfilling protection
  • Lid reduces the escaping vapours
  • Flame arrester made from stainless steel (simple to interchange)
  • Gevind:
    S 56×4
  • Beskrivelse:
    120 mm påfyldningsåbning