Safety funnels
120 mm påfyldningsåbning, S 56×4
DUEH5.10.001
Stainless steel safety funnels with hinged lid and butress thread S 56×4.
- Overfilling protection
- Lid reduces the escaping vapours
- Flame arrester made from stainless steel (simple to interchange)
- Gevind:S 56×4
- Beskrivelse:120 mm påfyldningsåbning