Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic bottle, 2,5 L
Katalog # 20910.330
Sundhed
Giftig
Ætsende
Leverandør:   VWR Chemicals
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Katalog # 20910.330
Leverandør:  VWR Chemicals
CAS nummer:  50-00-0
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic bottle, 2,5 L

CAS nummer:  50-00-0
Price
296,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS nummer:50-00-0
  • Formel:CH₂O
  • MDL nummer:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Tæthed:1,088 g/cm³ (20 °C)
  • Flash Pt:66 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-001-8
  • FN:2209
  • ADR:8,III
  •  :stabiliseret med methanol 10 %
Specifikationer

Specifikationstestresultater

  • Assay:
    35 - 38 %
  • Density (20/4):
    1.083 - 1.093
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Methanol:
    8 - 12 %

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    2,5 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.