Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Rolled rim vials

14,00 ml, 25,5×46 mm, klar, No
Katalog # MURRT103/V3
Leverandør:   MURRAY S
Rolled rim vials
Katalog # MURRT103/V3
Leverandør:  MURRAY S
Rolled rim vials
Rolled rim vials

14,00 ml, 25,5×46 mm, klar, No

Price
795,00 kr
Log ind for tilgængelighed
1 * 162 Items
Product Details & Documents

These vials without closure are made from neutral glass.

 : Polythene push-in plugs must be ordered separately.

Specifikationer
  • Kasket inkluderet:
    No
  • Diameter×H:
    25,5×46 mm
  • Kapacitet:
    14,00 ml
  • Farve:
    klar
  • Seq:
    02