Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rolled rim vials

Rolled rim vials

7,00 ml, 23×34 mm, klar, No
Katalog # 215-3570
Leverandør:   MURRAY S
Rolled rim vials
Katalog # 215-3570
Leverandør:  MURRAY S
Rolled rim vials
Rolled rim vials

7,00 ml, 23×34 mm, klar, No

Price
1.640,00 kr
Log ind for tilgængelighed
1 * 190 Items
Product Details & Documents

These vials without closure are made from neutral glass.

Ordering information: Polythene push-in plugs must be ordered separately.

Specifikationer
  • Kasket inkluderet:
    No
  • Diameter×H:
    23×34 mm
  • Kapacitet:
    7,00 ml
  • Farve:
    klar
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.