Brushes for test tubes
12 mm, Wire, 175 mm
Katalog # MURRL340/02
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Test tube brushes with bristles on wire handle.
Specifikationer
- Børstehoved Ø:12 mm
- Handle material:Wire
- Samlet længde:175 mm