Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Freeze drying ampoules

Freeze drying ampoules

Freeze drying ampoules, klar, 2,5 ml, 15×154 mm
Catalog # MURRG225/02
Supplier:   MURRAY S
Freeze drying ampoules
Catalog # MURRG225/02
Supplier:  MURRAY S
Freeze drying ampoules

Freeze drying ampoules, klar, 2,5 ml, 15×154 mm

Price
3.370,00 kr
Log in for availability
1 * 250 Items
Product Details & Documents

Freeze drying ampoules are made of neutral glass.

Specifications
  • Diameter×H:
    15×154 mm
  • Kapacitet:
    2,5 ml
  • Farve:
    klar
  • Beskrivelse:
    Freeze drying ampoules
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.