Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test tubes, without rim, medium wall, `SAMCO 50`

Test tubes, without rim, medium wall, `SAMCO 50`

125 mm, 16 mm, 1,2 mm
Katalog # 212-0457
Leverandør:   MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Katalog # 212-0457
Leverandør:  MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Test tubes, without rim, medium wall, `SAMCO 50`

125 mm, 16 mm, 1,2 mm

Price
780,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Test tubes, made of SAMCO neutral glas.

Neutral glass test tubes will withstand moderate temperature change, they should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.

Specifikationer
  • Udv.Ø:
    16 mm
  • Vægtykkelse:
    1,2 mm
  • Længde:
    125 mm
  • Seq:
    05
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.