Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test tubes, without rim, medium wall, `SAMCO 50`

Test tubes, without rim, medium wall, `SAMCO 50`

100 mm, 12 mm, 1,0 mm
Katalog # MURRG023/33
Leverandør:   MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Katalog # MURRG023/33
Leverandør:  MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Test tubes, without rim, medium wall, `SAMCO 50`

100 mm, 12 mm, 1,0 mm

Price
542,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Test tubes, made of SAMCO neutral glas.

Neutral glass test tubes will withstand moderate temperature change, they should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.

Specifikationer
  • Udv.Ø:
    12 mm
  • Vægtykkelse:
    1,0 mm
  • Længde:
    100 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.