Orders LinkContactUsLinkComponent
Test tubes, without rim, light wall
100 mm, 16 mm, 0,8 mm
  MURRG021/34
 :   MURRAY S
Test tubes, without rim, light wall
  MURRG021/34
 :  MURRAY S
Test tubes, without rim, light wall
100 mm, 16 mm, 0,8 mm
560,00 kr
1 * 100 Items
Glass, clear.
  • Excellent chemical resistance
  • Good temperature resistance
  • Udv.Ø:
    16 mm
  • Vægtykkelse:
    0,8 mm
  • Længde:
    100 mm