Funnels
200 mm
Katalog # 221-0165
Leverandør: NICKEL ELECTRO
Gå til:
Product Details & Documents
Rustfrit stål.
- Good chemical resistance, extremely high thermal resistance
- With air duct for discharging container air
- Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Specifikationer
- Tragt udv. Ø:200 mm
- Seq:03