Orders LinkContactUsLinkComponent
Dissection sets
Dissection kit, Basic Set in PVC roll, 9 pcs.
Catalog # 631-1092
Supplier:   MURRAY S
Dissection sets
Catalog # 631-1092
Supplier:  MURRAY S
Dissection sets
Dissection sets
Dissection kit, Basic Set in PVC roll, 9 pcs.
Price
330,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These high quality dissecting instruments are designed for everyday use. They are subject to stringent manufacturing quality procedures, and therefore suitable for surgical applications.
The Basic Set includes:
Two scalpels
Two needles
One seeker
One blunt forceps
One sharp forceps
One fine scissors
One blunt scissors
The Advanced Set includes:
Three scalpels
Two needles
One seeker
One section lifter
One blunt forceps
One sharp forceps
One sharp curved forceps
One fine scissors
One blunt scissors
One section razor
Specifications
  • Beskrivelse:
    Dissection kit, Basic Set in PVC roll, 9 pcs.