Dissecting scalpels
35 mm, Scalpel, solid forged, with polished blade, stainless steel
MURRD021/02
: MURRAY S
These solid forged scalpels are made from stainless steel and feature a polished blade.
- Beskrivelse:Scalpel, solid forged, with polished blade, stainless steel
- Klinge:35 mm