Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jenway® pH Electrodes

Jenway® pH Electrodes

Life science refillable combination with bulb end, pH, 0...80 °C, Glas, 12×120 mm, BNC, Jenway® pH meters
Katalog # JENW99960-41
Leverandør:   Cole-Parmer
Jenway® pH Electrodes
Katalog # JENW99960-41
Leverandør:  Cole-Parmer
Jenway® pH Electrodes
Jenway® pH Electrodes

Life science refillable combination with bulb end, pH, 0...80 °C, Glas, 12×120 mm, BNC, Jenway® pH meters

Price
1.630,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

A variety of pH electrodes for your Jewnay meters.

  • Choose the right electrode for your application

These electrodes are designed for standard measurements of general solutions. The epoxy body makes it especially suitable for field measurements or use in education.

Specifikationer
  • Tilslutning:
    BNC
  • Ø×L:
    12×120 mm
  • Til:
    Jenway® pH meters
  • Model:
    pH
  • Beskrivelse:
    Life science refillable combination with bulb end
  • Materiale:
    Glas
  • Seq:
    7
  • Temperatur:
    0...80 °C
  • Cat. No.:
    JENW99960-41
  • Kabellængde:
    1 m
  • Reference electrolyte:
    AgCl, liquid filled single junction
  • pH-område:
    0 - 14 pH
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.